2-Fluoro-3-formylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-3-formylbenzeneboronic acid, (2-Fluoro-3-formylphenyl)boronic acid

A162977-10G A162977-1G A162977-250MG A162977-5G A162977-25G
76985-128EA 41 USD
76985-128 76985-130 76985-132 76985-134 77890-069
2-Fluoro-3-formylphenylboronic acid 97%
2-Fluoro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MDL Number: MFCD07369725
CAS Number: 849061-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR