Val-Cit-PAB-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-[(S)-2-Amino-3-methylbutanamido]-N-[4-(hydroxymethyl)phenyl]-5-ureidopentanamide

A162959-100MG A162959-10MG A162959-1G A162959-1MG A162959-250MG A162959-50MG A162959-5G A162959-5MG
76985-112EA 62.6 USD
76985-112 76985-114 76985-116 76985-118 76985-120 76985-122 76985-124 76985-126
Val-Cit-PAB-OH 98%
Val-Cit-PAB-OH

Precaución: for Research Use Only

Formula: C₁₈H₂₉N₅O₄
MW: 379.46 g/mol
MDL Number: MFCD22381209
CAS Number: 159857-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR