L-Glutamic acid diethyl ester hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Diethyl L-glutamate, H-Glu(OEt)-OEt.HCl, H-Glu(OEt)-OEt·HCl

A162768-100G A162768-10G A162768-25G A162768-500G A162768-5G
76997-864EA 42 USD
76997-864 76997-866 76997-868 76997-870 76997-872
L-Glutamic acid diethyl ester hydrochloride 95%
L-Glutamic acid diethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₄
MDL Number: MFCD00012509
CAS Number: 1118-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR