2-Bromo-5-chloro-4-fluoronitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-chloro-5-fluoro-2-nitrobenzene

A162723-1G A162723-5G
76985-108EA 22.28 USD
76985-108 77611-588
2-Bromo-5-chloro-4-fluoronitrobenzene 97%
2-Bromo-5-chloro-4-fluoronitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFNO₂
MW: 254.441 g/mol
MDL Number: MFCD18089309
CAS Number: 960000-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR