5-(Trifluoromethyl)-1H-indazole 97%

Supplier: Ambeed
Warning

A162590-1G A162590-250MG A162590-5G
77011-278EA 28.65 USD
77011-278 77011-280 77011-282
5-(Trifluoromethyl)-1H-indazole 97%
5-(Trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
MDL Number: MFCD08234904
CAS Number: 885271-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR