5-Iodo-1,2,3-trimethylbenzene 96%

Supplier: Ambeed
Warning

A162495-100MG A162495-250MG A162495-1G
77011-270EA 57 USD
77011-270 77592-978 77890-066
5-Iodo-1,2,3-trimethylbenzene 96%
5-Iodo-1,2,3-trimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁I
MDL Number: MFCD27996326
CAS Number: 41381-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR