2-Phenylthiazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A162421-1G A162421-100MG
77011-268EA 182.48 USD
77011-268 77683-347
2-Phenylthiazole-5-carboxylic acid 97%
2-Phenylthiazole-5-carboxylic acid

Precaución: For research use only.

Formula: C₁₀H₇NO₂S
MDL Number: MFCD07376773
CAS Number: 10058-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR