5-Bromo-4-(tert-butyl)pyrimidine 97%

Supplier: Ambeed
Warning

A162355-100MG A162355-1G A162355-5G
76985-094EA 91.24 USD
76985-094 76985-096 76985-098
5-Bromo-4-(tert-butyl)pyrimidine 97%
5-Bromo-4-(tert-butyl)pyrimidine

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂
MW: 215.09 g/mol
MDL Number: MFCD17012062
CAS Number: 1439-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR