Methyl 5-bromo-4-chloronicotinate 97%

Supplier: Ambeed
Warning

A162328-100MG A162328-1G A162328-250MG
76985-090EA 91.24 USD
76985-090 76985-092 77683-343
Methyl 5-bromo-4-chloronicotinate 97%
Methyl 5-bromo-4-chloronicotinate

Precaución: For research use only.

Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
MDL Number: MFCD18257863
CAS Number: 1256808-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR