N-Boc-L-aspartic acid 4-cyclohexyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Asp(OcHex)-OH, 4-Cyclohexyl-N-(tert-butoxycarbonyl)-L-aspartate, Boc-Asp(OcHx)-OH, N-Boc-L-aspartic Acid 4-Cyclohexyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Cyclohexyl Ester, 4-Cyclohexyl N-Boc-L-aspartate

A162291-100G A162291-10G A162291-25G A162291-5G
77028-660EA 85 USD
77028-660 77028-662 77028-664 77028-666 77028-668
N-Boc-L-aspartic acid 4-cyclohexyl ester 97%
N-Boc-L-aspartic acid 4-cyclohexyl ester

Precaución: for Research Use Only

Formula: C₁₅H₂₅NO₆
MDL Number: MFCD00061996
CAS Number: 73821-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR