2-(3-Bromopyridin-2-yl)acetonitrile 98%

Supplier: Ambeed
Warning

A162147-1G A162147-250MG
76985-086EA 62 USD
76985-086 76985-088
2-(3-Bromopyridin-2-yl)acetonitrile 98%
2-(3-Bromopyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD09999977
CAS Number: 122851-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR