2-Chloro-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-pyridinol

A162114-1000G A162114-100G A162114-10G A162114-1G A162114-25G A162114-500G A162114-5G
77011-234EA 147 USD
77011-234 77011-236 77011-238 77011-240 77011-242 77011-244 77011-246
2-Chloro-3-hydroxypyridine 98%
2-Chloro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄ClNO
MDL Number: MFCD00006235
CAS Number: 6636-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR