3-Amino-2-hydroxy-N,N-dimethylbenzamide 95%

Supplier: Ambeed
Warning

A161910-100MG A161910-1G A161910-250MG
76985-080EA 45.62 USD
76985-080 76985-082 76985-084
3-Amino-2-hydroxy-N,N-dimethylbenzamide 95%
3-Amino-2-hydroxy-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
MDL Number: MFCD13191893
CAS Number: 464913-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR