3-(methylamino)benzonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Methylamino)benzonitrile

A161874-1G A161874-250MG
76985-076EA 98.67 USD
76985-076 76985-078
3-(methylamino)benzonitrile 95%
3-(methylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.163 g/mol
MDL Number: MFCD11153024
CAS Number: 64910-52-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR