4-Chloro-2-fluoronicotinaldehyde 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A161858-100MG A161858-250MG A161858-5G
76985-072EA 26 USD
76985-072 76985-074 77683-338
4-Chloro-2-fluoronicotinaldehyde 95%, Ambeed.Inc
4-Chloro-2-fluoronicotinaldehyde

Precaución: For research use only.

Formula: C₆H₃ClFNO
MW: 159.546 g/mol
MDL Number: MFCD15143389
CAS Number: 1232432-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR