(5-Chloro-2-fluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Chloro-2-fluorophenyl)acetic acid

A161834-1G A161834-10G A161834-25G
77028-654EA 14 USD
77028-654 77871-811 77871-812
(5-Chloro-2-fluorophenyl)acetic acid 97%
(5-Chloro-2-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MDL Number: MFCD01631439
CAS Number: 261762-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR