Adamantane-2,6-dione 97%

Supplier: Ambeed
Warning

A161748-1G A161748-250MG A161748-25G A161748-5G A161748-100MG
76985-064EA 48 USD
76985-064 76985-066 76985-068 76985-070 77890-059
Adamantane-2,6-dione 97%
Adamantane-2,6-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD01321093
CAS Number: 39751-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR