Folinic acid calcium salt 95%

Supplier: Ambeed
Warning

A161719-100MG A161719-1G A161719-1MG A161719-250MG A161719-25MG A161719-5G A161719-25G
76997-814EA 10 USD
76997-814 76997-816 76997-818 76997-820 76997-822 76997-824 77881-265
Folinic acid calcium salt 95%
Folinic acid calcium salt

Precaución: for Research Use Only

Formula: C₂₀H₂₁CaN₇O₇
MDL Number: MFCD00006704
CAS Number: 1492-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR