Methyl-4-chloropicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-chloropicolinate, Methyl 4-chloropyridine-2-carboxylate
A161562-100G
A161562-10G
A161562-500G
A161562-5G
A161562-25G
76997-798EA
56.23
USD
76997-798
76997-800
76997-802
76997-804
77319-732
Methyl-4-chloropicolinate 98%
Methyl 4-chloropicolinate
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.5800 g/mol |
MDL Number:
MFCD04116183 CAS Number: 24484-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...