Bis(4-(trifluoromethyl)phenyl)phosphine oxide 97%

Supplier: Ambeed
Warning

A161531-250MG A161531-5G A161531-1G
77011-218EA 178.24 USD
77011-218 77011-220 77567-356
Bis(4-(trifluoromethyl)phenyl)phosphine oxide 97%
Bis(4-(trifluoromethyl)phenyl)phosphine oxide

Precaución: For research use only.

Formula: C₁₄H₉F₆OP
MW: 338.19 g/mol
MDL Number: MFCD27952973
CAS Number: 15929-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR