2,6-Dihydroxybenzoic acid 97%

Supplier: Ambeed
Warning

A161205-1000G A161205-100G A161205-10G A161205-25G A161205-500G
76997-784EA 137 USD
76997-784 76997-786 76997-788 76997-790 76997-792
2,6-Dihydroxybenzoic acid 97%
2,6-Dihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₄
MDL Number: MFCD00002462
CAS Number: 303-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR