3-Bromo-6-(trifluoromethyl)-1H-indazole 98%

Supplier: Ambeed
Warning

A160986-100MG A160986-250MG
77011-192EA 36.08 USD
77011-192 77011-194
3-Bromo-6-(trifluoromethyl)-1H-indazole 98%
3-Bromo-6-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD09263228
CAS Number: 1000341-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR