5-Bromo-N-(furan-2-ylmethyl)pyrimidin-2-amine 98%

Supplier: Ambeed
Warning

A160591-1G A160591-5G
76985-036EA 42.44 USD
76985-036 76985-038
5-Bromo-N-(furan-2-ylmethyl)pyrimidin-2-amine 98%
5-Bromo-N-(furan-2-ylmethyl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₉H₈BrN₃O
MW: 254.08 g/mol
MDL Number: MFCD03646020
CAS Number: 886366-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR