4-Fluoro-2-indoleboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A160359-100MG A160359-1G A160359-250MG
77011-178EA 39 USD
77011-178 77011-180 77011-182
4-Fluoro-2-indoleboronic acid pinacol ester 98%
4-Fluoro-2-indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BFNO₂
MDL Number: MFCD16618972
CAS Number: 1256359-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR