Methyl-1-methyl-1H-indazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-indazole-4-carboxylate

A160333-100MG A160333-1G A160333-250MG
77028-614EA 14.86 USD
77028-614 77028-616 77028-618
Methyl-1-methyl-1H-indazole-4-carboxylate 95%
Methyl-1-methyl-1H-indazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.1986 g/mol
MDL Number: MFCD11109402
CAS Number: 1071428-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR