1-(1-Methyl-1H-imidazol-5-yl)ethanone 98%
Supplier: Ambeed
|
Warning
|
A160262-100MG
A160262-250MG
76985-028EA
48.81
USD
76985-028
76985-030
1-(1-Methyl-1H-imidazol-5-yl)ethanone 98%
1-(1-Methyl-1H-imidazol-5-yl)ethanone
Precaución: for Research Use Only
Formula:
C₆H₈N₂O MW: 124.14 g/mol |
MDL Number:
MFCD08668302 CAS Number: 20970-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...