Rhodanine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one

A160203-500G A160203-5G
76997-772EA 409.51 USD
76997-772 77592-952
Rhodanine 98%
Rhodanine

Precaución: for Research Use Only

Formula: C₃H₃NOS₂
MW: 133.19 g/mol
MDL Number: MFCD00005488
CAS Number: 141-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR