3-(3-Chloropropyl)-1,3-dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one 97%

Supplier: Ambeed
Warning

A160195-1G A160195-250MG A160195-25G A160195-5G
77028-604EA 7 USD
77028-604 77028-606 77028-608 77600-270
3-(3-Chloropropyl)-1,3-dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one 97%
3-(3-Chloropropyl)-1,3-dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₈ClNO₃
MDL Number: MFCD09955404
CAS Number: 85175-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR