2-(Benzylamino)nicotinic acid 98%

Supplier: Ambeed
Warning

A160187-250MG A160187-5G
76997-768EA 9.55 USD
76997-768 76997-770 77881-252
2-(Benzylamino)nicotinic acid 98%
2-(Benzylamino)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₂
MDL Number: MFCD01568566
CAS Number: 33522-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR