tert-Butyl (3-aminocyclopentyl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

A160159-100MG A160159-10G A160159-1G A160159-5G
77028-596EA 56 USD
77028-596 77028-598 77028-600 77028-602
tert-Butyl (3-aminocyclopentyl)carbamate hydrochloride 97%
tert-Butyl (3-aminocyclopentyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD09759162
CAS Number: 1197239-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR