2,6-Dibromo-4-hydroxybenzonitrile 95%
Supplier: Ambeed
|
Warning
|
A160088-100MG
A160088-1G
A160088-250MG
76985-020EA
22
USD
76985-020
76985-022
76985-024
2,6-Dibromo-4-hydroxybenzonitrile 95%
2,6-Dibromo-4-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃Br₂NO |
MDL Number:
MFCD07787512 CAS Number: 74283-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...