BAPTA tetraethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Tetraethyl 2,2',2'',2'''-(((ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetate

A159884-1G A159884-250MG A159884-5G
76997-748EA 71.09 USD
76997-748 76997-750 76997-752
BAPTA tetraethyl ester 98%
BAPTA tetraethyl ester

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₂O₁₀
MW: 588.66 g/mol
MDL Number: MFCD00532663
CAS Number: 73630-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR