3-Nitro-2,6-lutidine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,6-Dimethyl-3-nitropyridine, 3-Nitro-2,6-lutidine
A159756-10G
A159756-1G
A159756-5G
76997-742EA
30.77
USD
76997-742
76997-744
76997-746
3-Nitro-2,6-lutidine 98%
3-Nitro-2,6-lutidine
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol |
MDL Number:
MFCD00023503 CAS Number: 15513-52-7 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...