3-Methyl-2-(methylsulfonamido)benzoic acid 95%

Supplier: Ambeed
Warning

A159696-1G A159696-250MG
77011-160EA 168.69 USD
77011-160 77011-162
3-Methyl-2-(methylsulfonamido)benzoic acid 95%
3-Methyl-2-(methylsulfonamido)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄S
MW: 229.253 g/mol
MDL Number: MFCD09045450
CAS Number: 926206-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR