Fmoc-Lys(ipr,Boc)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nepsilon-Boc-Nalpha-Fmoc-Nepsilon-isopropyl-L-lysine, N-Fmoc-N'-boc-N'-isopropyl-L-Lysine, Fmoc-Lys(Boc)(isopropyl)-OH
A159686-1G
A159686-250MG
A159686-25G
A159686-5G
76985-008EA
33
USD
76985-008
76985-010
76985-012
76985-014
Fmoc-Lys(ipr,Boc)-OH 95%
Fmoc-Lys(ipr,Boc)-OH
Precaución: for Research Use Only
Formula:
C₂₉H₃₈N₂O₆ MW: 510.631 g/mol |
MDL Number:
MFCD00080276 CAS Number: 201003-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...