Bis(cyclopentadienyl)titanium dichloride 98%
Supplier: Ambeed
|
Warning
|
A159684-25G
A159684-5G
A159684-100G
A159684-500G
76997-738EA
15.92
USD
76997-738
76997-740
77550-006
77550-008
Bis(cyclopentadienyl)titanium dichloride 98%
Bis(cyclopentadienyl)titanium dichloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₀Cl₂Ti |
MDL Number:
MFCD00003723 CAS Number: 1271-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...