3-Amino-5-chloro-1H-indazole 98%

Supplier: Ambeed
Warning

A159389-100MG A159389-1G A159389-250MG
77028-578EA 22.28 USD
77028-578 77028-580 77028-582
3-Amino-5-chloro-1H-indazole 98%
3-Amino-5-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.60 g/mol
MDL Number: MFCD07385612
CAS Number: 5685-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR