4-Chloroquinolin-5-amine 95%

Supplier: Ambeed
Danger

A159373-1G A159373-100MG A159373-250MG
77011-138EA 256 USD
77011-138 77611-536 77611-538
4-Chloroquinolin-5-amine 95%
4-Chloroquinolin-5-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MDL Number: MFCD20259272
CAS Number: 1824318-74-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR