2-Bromo-3,5-dichloro-4-methylpyridine 98%

Supplier: Ambeed
Warning

A159349-100MG A159349-1G A159349-250MG A159349-5G
76984-998EA 40.32 USD
76984-998 76985-000 76985-002 76985-004
2-Bromo-3,5-dichloro-4-methylpyridine 98%
2-Bromo-3,5-dichloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrCl₂N
MW: 240.91 g/mol
MDL Number: MFCD26383615
CAS Number: 344324-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR