(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, (1S,2S)-2-(Diphenylphosphanyl)-1,2-diphenylethan-1-amine, [(1S,2S)-1,2-Diphenyl-2-aminoethyl]diphenylphosphine, (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethanamine

A159300-100MG A159300-1G A159300-250MG
76984-992EA 102.91 USD
76984-992 76984-994 76984-996
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine ≥97%, ee 99%
(S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine

Precaución: for Research Use Only

Formula: C₂₆H₂₄NP
MW: 381.459 g/mol
MDL Number: MFCD11045444
CAS Number: 1091606-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR