6-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 95+%

Supplier: Ambeed
Warning

A159281-100MG A159281-1G A159281-250MG
76984-986EA 62 USD
76984-986 76984-988 76984-990
6-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 95+%
6-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.09 g/mol
MDL Number: MFCD16619138
CAS Number: 6729-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR