Fmoc-D-CIS-HYP-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-cis-Hyp-OH

A159263-10G A159263-1G A159263-250MG A159263-5G
76997-714EA 349.04 USD
76997-714 76997-716 76997-718 76997-720
Fmoc-D-CIS-HYP-OH 97%
Fmoc-D-CIS-HYP-OH

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₅
MW: 353.374 g/mol
MDL Number: MFCD01318735
CAS Number: 214852-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR