Cbz-Ser(Bzl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Z-Ser(Bzl)-OH, N-Benzyloxycarbonyl-O-benzyl-L-serine

A159135-100G A159135-10G A159135-1G A159135-250MG
76997-706EA 271.6 USD
76997-706 76997-708 76997-710 76997-712
Cbz-Ser(Bzl)-OH 95%
Cbz-Ser(Bzl)-OH

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₅
MW: 329.4 g/mol
MDL Number: MFCD00022029
CAS Number: 20806-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR