2-Bromo-6-fluorobenzothiazole 95%

Supplier: Ambeed
Warning

A159088-1G A159088-5G
76997-702EA 22.28 USD
76997-702 76997-704
2-Bromo-6-fluorobenzothiazole 95%
2-Bromo-6-fluorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₃BrFNS
MDL Number: MFCD07783786
CAS Number: 152937-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR