Methyl 3-bromo-2-chlorobenzoate 95%

Supplier: Ambeed
Warning

A158896-10G A158896-1G A158896-250MG A158896-5G
77011-118EA 127 USD
77011-118 77011-120 77011-122 77011-124
Methyl 3-bromo-2-chlorobenzoate 95%
Methyl 3-bromo-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD18398720
CAS Number: 871224-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR