Oxindole-6-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Oxoindolin-6-yl)boronic acid

A158860-100MG A158860-1G A158860-5G A158860-250MG
76984-966EA 51.99 USD
76984-966 76984-968 76984-970 77683-324
Oxindole-6-boronic acid 97%
Oxindole-6-boronic acid

Precaución: For research use only.

Formula: C₈H₈BNO₃
MW: 176.9650 g/mol
MDL Number: MFCD12913936
CAS Number: 1217500-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR