1-(2-Amino-6-fluorophenyl)ethanone 98%

Supplier: Ambeed
Warning

A158784-100MG A158784-1G A158784-250MG
77011-110EA 75.33 USD
77011-110 77011-112 77011-114
1-(2-Amino-6-fluorophenyl)ethanone 98%
1-(2-Amino-6-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.15 g/mol
MDL Number: MFCD12025288
CAS Number: 869937-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR