Oxyresveratrol 98%

Supplier: Ambeed
Warning

Synonyms: 2,3',4,5'-Tetrahydroxy-trans-stilbene, (E)-4-(3,5-Dihydroxystyryl)benzene-1,3-diol

A158758-1G A158758-250MG A158758-50MG A158758-5G A158758-10MG A158758-100G A158758-100MG A158758-25G
77028-552EA 25 USD
77028-552 77028-554 77028-556 77028-558 77667-236 77871-791 77871-792 77871-793
Oxyresveratrol 98%
Oxyresveratrol

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MDL Number: MFCD11974969
CAS Number: 29700-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR