5-Chloro-3-fluoro-2-hydroxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-fluoropyridin-2-ol

A158712-1G A158712-250MG A158712-5G A158712-25G
77011-102EA 20 USD
77011-102 77011-104 77011-106 77890-039
5-Chloro-3-fluoro-2-hydroxypyridine 97%
5-Chloro-3-fluoro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFNO
MDL Number: MFCD07375044
CAS Number: 514797-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR