6-Methylpyrazine-2-carbonitrile 95%

Supplier: Ambeed
Warning

A158662-100MG A158662-1G A158662-250MG
76984-956EA 46 USD
76984-956 76984-958 76984-960
6-Methylpyrazine-2-carbonitrile 95%
6-Methylpyrazine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.12 g/mol
MDL Number: MFCD08705762
CAS Number: 136309-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR